Thymine 99%
Plastic bottle, 500 g
Fournisseur: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
- CAS Number:65-71-4
- Formula:C₅H₆N₂O₂
- MDL Number:MFCD00006026
- Molecular Weight:126.11 g/mol
- Merck Index:12,09539
- Boiling Pt:389 °C (1013 hPa)
- Melting Pt:316…317 °C
- Density:1,454 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:200-616-1
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.01 %
- Calcium (Ca):≤0.002 %
- Iron (Fe):≤0.002 %
- Sodium (Na):≤0.005 %
- Insoluble matter:≤0.005 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g



