Thymine 99%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
- CAS Number:65-71-4
- Formula:C₅H₆N₂O₂
- MDL Number:MFCD00006026
- Molecular Weight:126.11 g/mol
- Merck Index:12,09539
- Boiling Pt:389 °C (1013 hPa)
- Melting Pt:316…317 °C
- Density:1,454 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:200-616-1
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.01 %
- Calcium (Ca):≤0.002 %
- Iron (Fe):≤0.002 %
- Sodium (Na):≤0.005 %
- Insoluble matter:≤0.005 %
Specifications
- Pack type:Glass bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



