Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanillin 99%, pure

Vanillin 99%, pure

Plastic drum, 5 kg
Catalog # ACRO140820050
Irritant
Vanillin 99%, pure
Catalog # ACRO140820050
CAS Number:  121-33-5
Vanillin 99%, pure
Vanillin 99%, pure

Plastic drum, 5 kg

CAS Number:  121-33-5
Price
571,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:170 °C (15 mmHg)
  • Melting Pt:81…84 °C
  • Density:1,056 g/cm³ (20 °C)
  • Flash Pt:147 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    81 to 84 °C
  • Titration with NaOH:
    ≥98.5 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 kg

Specifications