Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium deoxycholate 99%, extra pure

Sodium deoxycholate 99%, extra pure

Glass bottle, 500 g
Catalog # ACRO218595000
Irritant
Sodium deoxycholate 99%, extra pure
Catalog # ACRO218595000
CAS Number:  302-95-4
Sodium deoxycholate 99%, extra pure
Sodium deoxycholate 99%, extra pure

Glass bottle, 500 g

CAS Number:  302-95-4
Price
348,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  • CAS Number:302-95-4
  • Formula:C₂₄H₃₉NaO₄
  • MDL Number:MFCD00064139
  • Molecular Weight:414.56 g/mol
  • Melting Pt:357…365 °C
  • Storage Temperature:Ambient
  • EINECS:206-132-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White to cream Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5 % (2 g, 105°C, 4 h)
  • TLC:
    ≤2 % Sodium cholate
  • pH:
    7.5 to 10 (5 % in water)
  • Specific optical rotation:
    +42° to +46° (20°C, 589 nm) (c=2, water)
  • Titration with HClO4:
    98.5 to 102.0 % (on dry substance)
  • Heavy metals (as Pb):
    ≤20 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.