Buffer solution pH 10, carbonate buffer
10.00, ±0,02, Carbonate, Colourless, Plastic bottle, 2,5 L
Fournisseur: Thermo Fisher Scientific
Buffer solution pH 10, carbonate buffer
N° de catalogue ACRO258600025
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Aller à :
Product Details & Documents
CAS No.: 1332-57-6
MDL No.: MFCD00036143
Appearance: Clear colourless solution
Storage Temperature: Ambient
Documents
Spécifications
Spécifications
- Pack type:Plastic bottle
- pH buffer type:Carbonate
- Pk:2,5 L
- Colour:Colourless
- pH accuracy:±0,02
- pH value:10.00



