Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solution pH 7, Phosphate buffer

Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution, 7,00, ±0,02, Phosphate, Plastic bottle, 2,5 L
Catalog # ACRO258590025
Buffer solution pH 7, Phosphate buffer
Catalog # ACRO258590025
CAS Number:  
Buffer solution pH 7, Phosphate buffer

Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution, 7,00, ±0,02, Phosphate, Plastic bottle, 2,5 L

CAS Number:  
Price
83,80 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Phosphate
  • Pk:
    2,5 L
  • Description:
    Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
  • pH accuracy:
    ±0,02
  • pH value:
    7,00
  • Cat. No.:
    ACRO258590025
  • Concentration:
    3.54 g potassium dihydrogen phosphate and 14.7 g i-sodium hydrogen phosphate per litre
  • Appearance:
    Clear colourless solution

Specifications