Buffer solution pH 4, citrate buffer
4,00, ±0,02, Citrate, Colourless, Plastic bottle, 5 L
Documents
ACRO25858
SDS
- Pack type:Plastic bottle
- pH buffer type:Citrate
- Pk:5 L
- Colour:Colourless
- pH accuracy:±0,02
- pH value:4,00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



