Galactitol 99+%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:608-66-2
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064288
- Molecular Weight:182.17 g/mol
- Boiling Pt:275…280 °C (1 mmHg)
- Melting Pt:188…189 °C
- Storage Temperature:Ambient
- EINECS:210-165-2
Documents
ACRO11770
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:187 to 191 °C
- Water:≤1.0 % (K.F.)
- Solubility:(3 % in water) Colorless
- HPLC:≥99.0 % (ELSD)
- Heavy metals (as Pb):≤0.003 %
- Insoluble matter:≤0.01 % (in dilute NH4OH)
Specifications
- Pack type:Glass bottle
- Pk:500 g



