ContactUsLinkComponent
B ClearKlensIPA sterilIPA70%300ml7514935Sterile disinfectant, aerosol, 70% Isopropyl alcohol and30% Water(purified water), 0,2µm fil tered, gammairradiated, double bagged, 300 ml aerosol bottle,packaging unit 12 x 300 ml 1 * 12 Bottle
Catalog # DIVY7514935
Supplier:  Diversey
B ClearKlensIPA sterilIPA70%300ml7514935Sterile disinfectant, aerosol, 70% Isopropyl alcohol and30% Water(purified water), 0,2µm fil tered, gammairradiated, double bagged, 300 ml aerosol bottle,packaging unit 12 x 300 ml 1 * 12 Bottle
Catalog # DIVY7514935
Supplier:  Diversey
Frequently Bought Together