Orders LinkContactUsLinkComponent
VWR® Overalls, SMS
VWR® Overalls, SMS
White, M
Catalog # 113-8226
Supplier:   VWR Collection
VWR® Overalls, SMS
Catalog # 113-8226
Supplier:  VWR Collection
VWR® Overalls, SMS
VWR® Overalls, SMS
White, M
Price
404,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
These durable, anti-static overalls are made of breathable, soft SMS, a spunbonded fabric with a unique tri-laminate construction, that offers a high tensile strength and toughness, plus provides a barrier to fluids and particulates.
  • Elasticated hood
  • Zip closure with flap
  • Elasticated cuffs, ankles and waist, thumb loop
  • Serged seams (stitching method)
  • Low lint performance
These overalls can be used in medical, industrial, laboratory, electronic and pharmaceutical applications. Suitable for use in Critical Environments (IEST-RP-CC003.3 – Level I) and ISO Class 5 + 6 cleanrooms.
Weight: 60 g/m²
EN 340, EN 1149-5, EN 13034:2005+A1:2009 (Type 5), EN 13982-1:2004+A1:2010 (Type 6), EN 1073-2:2002; Cat. III
Certifications: EN 340, EN 1149-5, EN 13034+A1 (Type 5), EN 13982-1+A1 (Type 6), EN 1073-2; Cat. III
Specifications
  • Clothing Size:
    M
  • Colour:
    White
  • Seq:
    02
  • Cat. No.:
    113-8226
  • Anti-static:
    +
  • Garment application:
    Cleanroom|Medical|Laboratory|Clinical|Industrial
  • Weight:
    60 g/m²