Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample tubes

Sample tubes

30 ml, 25,3 mm, 111 mm, White, sterile
  SIMPC572-1
Sample tubes
  SIMPC572-1
Sample tubes
Sample tubes

30 ml, 25,3 mm, 111 mm, White, sterile

 
261,00 €
   
1 * 500 ST
 

These tubes are chemically resistant and shatterproof, they are supplied with a leakproof screw cap, particularly important when transporting hazardous material.

  • Tube made of PP
  • Cap made of PE
  • Self-standing base with graduation
  • Leakproof screw cap
  • Available sterile and non-sterile
  • Functional temperature range: –90 to +121 °C

Conical bottom tubes with self-standing base graduated from 10 to 30 ml and 10 to 50 ml respectively in 5 ml increments. Ideal for transport and storage of urine, sputum and most liquids or particulate samples. Molded-in graduations make them easy to read.

Packaging: Please note that the non-sterile caps and tubes are package separately in packages of 100. Sterile tubes are supplied in packs of 25.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

 
  • Ext.Ø:
    25,3 mm
  • Version:
    sterile
  • Capacity:
    30 ml
  • Height:
    111 mm
  • Cap colour:
    White
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.