Accessories
Discharge tube, with recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
Catalog # 613-5217
Supplier: Brand
Jump to:
Product Details & Documents
Documents
Specifications
- Description:Discharge tube, with recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
- Seq:17