ContactUsLinkComponent
Discharge valve for Dispensette® S 1 and 2 ml, PFA/glass/ceramic/platinum-iridium, valve marked with 1 + 2 1 * 1 items
Catalog # BRND6749
Supplier:  Brand
undefined
Discharge valve for Dispensette® S 1 and 2 ml, PFA/glass/ceramic/platinum-iridium, valve marked with 1 + 2 1 * 1 items
Catalog # BRND6749
Supplier:  Brand