Discharge valve for Dispensette® S 5 and 10 ml, PFA/glass/ceramic/platinum-iridium, no valve marking 1 * 1 items
Catalog # BRND6727
Supplier: Brand
Discharge valve for Dispensette® S 5 and 10 ml, PFA/glass/ceramic/platinum-iridium, no valve marking 1 * 1 items
Catalog # BRND6727
Supplier: Brand
Frequently Bought Together
Bunsen burners, LABOGAZ 206
Supplier: CAMPING GAZ
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for PIPETBOY acu / pro
Supplier: Integra
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Accessories for accu-jet® pro
Supplier: Brand
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Iodine 0.025 M (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
AGAR - PLATES 90MM DG 18 1 * 20 items
Supplier: LIOFILCHEM
Accessories for water purification millipore
Supplier: Merck
Calibration software, EASYCAL™ 5.0
Supplier: Brand