Order Entry
Belgium
Orders LinkContactUsLinkComponent
Push button lockout devices

Push button lockout devices

Medium (for 22 mm push buttons), Red
Catalog # BRDY130819
Supplier:   Brady
Push button lockout devices
Catalog # BRDY130819
Supplier:  Brady
Push button lockout devices
Push button lockout devices

Medium (for 22 mm push buttons), Red

Price
30,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

These lockout devices prevent the unauthorised use of push button emergency devices.

  • With standard cover
  • Made from clear PC, with translucent red cover
  • Suitable for 16, 22 or 30 mm push buttons
Specifications
  • Type:
    Medium (for 22 mm push buttons)
  • Colour:
    Red
  • Seq:
    04