Orders LinkContactUsLinkComponent
Inoculating loops, PS, calibrated
Inoculating loop, PS, sterile, 1 µl, Green
N° de catalogue DELT302743
Fournisseur:   DELTALAB
Inoculating loops, PS, calibrated
N° de catalogue DELT302743
Fournisseur:  DELTALAB
Inoculating loops, PS, calibrated
Inoculating loop, PS, sterile, 1 µl, Green
Prix
186,00 €
Log in for availability
1 * 1.200 ST
Product Details & Documents
These calibrated, dual use inoculating loops offer a loop at one end to facilitate microbiological streaking, and a fine tip at the other specially designed for counting colonies.
  • High robustness and flexible
  • Colour-coded for ease of size identification
  • Eliminates the need for flaming
  • Sterilised by radiation
Specifications
  • Capacity:
    1 µl
  • Colour:
    Green
  • Description:
    Inoculating loop, PS, sterile