Lead(II) acetate USP Test Solution (TS)
Plastic bottle, 100 mL
Supplier: VWR Chemicals
Lead(II) acetate USP Test Solution (TS)
Catalog # 85351.180
Supplier: VWR Chemicals
CAS Number: 301-04-2
Jump to:
Product Details & Documents
Ready to use test solution (TS).
- Produced according to Pharmacopoeia requirements
- Reduces time and expenses
- Independent, traceable and certified
- Conforms to USP 34
- Certificates of analysis and safety data sheets available
Synonyms: Acetic acid Lead(II) salt
- CAS Number:301-04-2
- Formula:(H₃CCOO)₂Pb
- MDL Number:MFCD00012452
- Molecular Weight:325.29 g/mol
- Merck Index:12,05411
- Melting Pt:280 °C
- EINECS:0
- UN:1616
- ADR:6.1,III
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:100 mL



