ContactUsLinkComponent
High Precision Streptavidin (SAX) Biosensors - One tray of 96 precision-controlled (CV<4%) streptavidin biosensors for high precision quantitation, screening, or kinetic applications. 1 * 1 items
Catalog # PALF18-5117
Supplier:  PALL FORTEBIO
High Precision Streptavidin (SAX) Biosensors - One tray of 96 precision-controlled (CV<4%) streptavidin biosensors for high precision quantitation, screening, or kinetic applications. 1 * 1 items
Catalog # PALF18-5117
Supplier:  PALL FORTEBIO