Closures
Screw cap with PTFE insert, GL 45, Black, PTFE
Catalog # RIXIU-8S-100445100
Supplier: BERLIN PACKAGING RIXIUS GMBH
Aller à :
Product Details & Documents
Spécifications
- Thread:GL 45
- Colour:Black
- Description:Screw cap with PTFE insert
- Material:PTFE
- Seq:15