Wall cleaning tool
Mopp MicroSicuro CR/A for wall cleaning tool (160 g): Special multiple-use cleanroom mop, for the cleaning of walls, roofs and difficult to access areas, safe disinfection of GMP areas through vailidatable moistening (100 % PES-microfiber with special soft bristle), 380×70 mm
Catalog # PPSA3580039
Supplier: PFENNIG REINIGUNGSTECHNIK
Jump to:
Product Details & Documents
Wall cleaning tool for wall and ceiling cleaning in hygienically sensitve areas.
- Frame completely made of stainless steel
- Mops are 100% polyester microfiber filament yarn
- Autoclavable, disinfectant proof
- Mops have high-quality stainless steel push buttons for fitting with frame
This mop frame with locking screw is suitable with mop covers Micro Sicuro CR/A and Mop CR for wall cleaning tool.
Packaging: Mops are prewashed and single double packaged.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Mopp MicroSicuro CR/A for wall cleaning tool (160 g): Special multiple-use cleanroom mop, for the cleaning of walls, roofs and difficult to access areas, safe disinfection of GMP areas through vailidatable moistening (100 % PES-microfiber with special soft bristle)
- L×W:380×70 mm
- Seq:03
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



