Orders LinkContactUsLinkComponent
Wall cleaning tool
Wall cleaning tool
Wall cleaning tool stainless steel: For wall and ceiling cleaning, with handle pick-up made of stainless steel, - mm
Catalog # PPSA3580033
Wall cleaning tool
Catalog # PPSA3580033
Wall cleaning tool
Wall cleaning tool
Wall cleaning tool stainless steel: For wall and ceiling cleaning, with handle pick-up made of stainless steel, - mm
Price
98,45 €
Log in for availability
1 * 1 ST
Product Details & Documents
Wall cleaning tool for wall and ceiling cleaning in hygienically sensitve areas.
  • Frame completely made of stainless steel
  • Mops are 100% polyester microfiber filament yarn
  • Autoclavable, disinfectant proof
  • Mops have high-quality stainless steel push buttons for fitting with frame
This mop frame with locking screw is suitable with mop covers Micro Sicuro CR/A and Mop CR for wall cleaning tool.
Packaging: Mops are prewashed and single double packaged.
Documents
Specifications
  • Description:
    Wall cleaning tool stainless steel: For wall and ceiling cleaning, with handle pick-up made of stainless steel
  • L×W:
    - mm
  • Seq:
    01
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.