Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Immunoassay Blocking Buffer

Immunoassay Blocking Buffer

Immunoassay blocking buffer, pre-coated, 50 ml
  ABCAAB171534-50
Marketsource
 :   Abcam
Immunoassay Blocking Buffer
  ABCAAB171534-50
 :  Abcam
Marketsource
 :  
Immunoassay Blocking Buffer
Immunoassay Blocking Buffer

Immunoassay blocking buffer, pre-coated, 50 ml

 :  
 
82,00 €
   
1 * 50 mL
 

Immunoassay blocking buffer effectively preserves the conformation and activity of dried proteins in immunoassays.

Caution: RUO.

 

 

  • Désignation:
    Immunoassay blocking buffer, pre-coated
  • Volume:
    50 ml
  • Cat. No.:
    ABCAAB171534-50
  • Température de stockage:
    Store at room temperature

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.