Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Filter discs, DURAN®

0, 55 mm, 5 mm
  SCOT251555005
Filter discs, DURAN®
  SCOT251555005
Filter discs, DURAN®
Filter discs, DURAN®

0, 55 mm, 5 mm

 
18,80 €
   
1 * 1 ST
 

Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.

  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design

Made from clear DURAN® borosilicate glass 3,3.

 
  • Ø:
    55 mm
  • Porosity:
    0
  • Seq:
    10
  • Thickness:
    5 mm