BD® Blunt Needles
18 G, 25 mm, Blunt bevel, Sterile, Red
BDAM305181
BD blunt fill and blunt filter needles are specifically designed to reduce the risk of needlestick injuries during medication preparation.
- The blunt fill needle is designed for vial access and available with a BD Luer-Lok™ connection
- The blunt filter is designed for safe ampoule medication preparation
- BD blunt fill needles have female luer fittings, which mate to male luer fittings
- BD blunt filter needle has a 5-micron filter for particles such as glass and plastic fragments, for safer medication delivery
- Natural rubber latex is not part of the material formulation
- Sterile, singe use
Caution: Note: The BD blunt fill and filter needles are not for skin injection.
- Colour code:Red
- Gauge:18 G
- Type:Sterile
- Type of tip:Blunt bevel
- Length:25 mm
- Seq:02