Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH buffer solutions, colour coded

7,00, ±0,01, Yellow, 25 °C, 500 mL
  REGE1070525C
 :   REAGECON
pH buffer solutions, colour coded
  REGE1070525C
 :  REAGECON
 :  
pH buffer solutions, colour coded

7,00, ±0,01, Yellow, 25 °C, 500 mL

 :  
 
38,40 €
   
1 * 500 mL
 

Colour coded pH buffer solutions for the calibration of pH meters.

  • Can be tested at 20 or 25 °C

 : Certified by Reagecon’s ISO 17025 accredited test method and NIST traceable.

 

 

 

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Yellow
  • pH accuracy:
    ±0,01
  • pH value:
    7,00