Orders LinkContactUsLinkComponent
Mini-pictograms
Mini-pictograms
Hazard labels, Mini 2 mandatory set, MINI-2, Vinyl, White/blue, 96 labels/sheet
Catalogus # BRDY286905
Leverancier:   Brady
Mini-pictograms
Catalogus # BRDY286905
Leverancier:  Brady
Mini-pictograms
Mini-pictograms
Hazard labels, Mini 2 mandatory set, MINI-2, Vinyl, White/blue, 96 labels/sheet
Price
10,20 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These labels are made from transparent vinyl film with permanent adhesive and are specifically designed to be put on your emergency plans.
Specificaties
  • Colour:
    White/blue
  • Description:
    Hazard labels, Mini 2 mandatory set
  • Material:
    Vinyl
  • Seq:
    11
  • Model:
    MINI-2
  • Packed:
    96 labels/sheet