Osmolality standards
Product Details & Documents
Osmolality standards provide ease of use when calibrating all types of osmometers. All Osmolality standards are manufactured using high purity raw materials. These products are prepared gravimetrically on a weight/weight basis. Both solute (salts) and solvent (water) are weighed on a balance calibrated by using OIML traceable weights.
- Ready to use and presented in convenient ampoules
- Extended shelf life and has low uncertainty of measurement
- Consistency of product-Independent, traceable
- Consist of 12 ampoules of 5 ml
The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.
Certifications: ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
Documents
Specifications
Specifications
- Pack type:Ampoule
- Pk:1 KIT
- Description:Osmolality standard
- Pk Info:12×5 ml
- Osmolality:1000 mOsm/kg H₂O