Mobile container spill kit
Mobile container maintanence kit, 124 L, 500×540×900 mm
BRDY813882
: Brady
These wheeled spill kits are comes in a polyethylene container with hinged lid. Designed for medium sized spills of industrial liquids including oils, water, solvents, coolants and chemical hazardous liquids.
- Yellow in colour
Container is weather-resistant and protects the sorbents against dirt, moisture and damage. Hinged lid provides easy access to the products. Wheels allow excellent mobility indoors and outdoors.
- Désignation:Mobile container maintanence kit
- Seq:01
- l×p×h:500×540×900 mm
- Capacité d'absorption:124 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



