Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
200 ml, 14/15, 0,15 ± ml
Catalog # POULRB258/H/2
Supplier:   Poulten & Graf
Volumetric flasks, class A
Catalog # POULRB258/H/2
Supplier:  Poulten & Graf
Volumetric flasks, class A
Volumetric flasks, class A
200 ml, 14/15, 0,15 ± ml
Price
47,70 €
Log in for availability
1 * 2 ST
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
Specifications
  • NS:
    14/15
  • Capacity:
    200 ml
  • Seq:
    25
  • Tolerance:
    0,15 ± ml