Order Entry
Belgium
Orders LinkContactUsLinkComponent
Culture media, ready to use, in plates

Culture media, ready to use, in plates

Rambach agar, -, 20 ml, 20
Catalog # 710-2267
Supplier:   MERCK LIFE SCIENCE
Culture media, ready to use, in plates
Catalog # 710-2267
Supplier:  MERCK LIFE SCIENCE
CAS Number:  
Culture media, ready to use, in plates
Culture media, ready to use, in plates

Rambach agar, -, 20 ml, 20

CAS Number:  
Price
87,60 €
Log in for availability
1 * 20 ST
Product Details & Documents

The product range includes a full line of pre-poured 90 mm or 150 mm Petri dishes for a wide range of different applications, such as enumeration, isolation, detection and cultivation of bacteria as well as yeast and moulds. In order to choose the best packaging for each kind of use, both small packs of 20 plates and bulk packs of 100 plates are available for each kind of plate.

  • Ready to use media in 90 mm Petri dishes
  • Item number and the expiry date are imprinted on every plate for safe and easy product identification and shelf life monitoring

Packaging: 2 or 10 sets of 10 shrink-wrapped plates

Specifications

Specifications

  • Pk:
    20
  • Standards:
    -
  • Description:
    Rambach agar
  • Filling:
    20 ml

Specifications

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Rack, stainless steel, 240 mm, for 10× Ø 90 mm settle plates
    Rack, stainless steel, 240 mm, for 10× Ø 90 mm settle plates
    HEIP1465190001
    Log in for availability
    1 * 1 ST - 102,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.