Orders LinkContactUsLinkComponent
Bottle racks
Bottle racks
Bottle rack, 6× 1 litre (max. 100 mm diameter)
Catalog # HEIP1465250001
Supplier:   MERCK LIFE SCIENCE
Bottle racks
Catalog # HEIP1465250001
Supplier:  MERCK LIFE SCIENCE
Bottle racks
Bottle rack, 6× 1 litre (max. 100 mm diameter)
Price
219,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Racks for media bottles, made of high quality stainless steel, electrolytic polished.
  • Autoclavable
  • Easy to clean
  • Good chemical and temperature resistance
Safe and convenient transport of bottles. Suitable for isolators and clean rooms.
Specifications
  • Description:
    Bottle rack, 6× 1 litre (max. 100 mm diameter)
  • Seq:
    02