Media in Bottles
Yeast glucose Chloramphenicol agar, —, 200 ml, Without cap, 250 ml, 6
Supplier: MERCK LIFE SCIENCE
Jump to:
Product Details & Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:6
- Standards:—
- Description:Yeast glucose Chloramphenicol agar
- Size:250 ml
- Closure type:Without cap
- Filling:200 ml



