Media in Bottles
Milk Plate Count Agar, —, 200 ml, Without cap, 250 ml, 6
- Pk:6
- Standards:—
- Description:Milk Plate Count Agar
- Size:250 ml
- Closure type:Without cap
- Filling:200 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



