Media in Bottles
MRS agar pH 5,4, —, 200 ml, Without cap, 250 ml, 6
Catalog # HEIP1463640006
Supplier: MERCK LIFE SCIENCE
Jump to:
Product Details & Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:6
- Standards:—
- Description:MRS agar pH 5,4
- Size:250 ml
- Closure type:Without cap
- Filling:200 ml



