Media in Bottles
Thioglycollate broth, EP, USP, 100 ml, Flip cap, 125 ml, 10
Catalog # HEIP1463330010
Supplier: MERCK LIFE SCIENCE
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:10
- Standards:EP, USP
- Description:Thioglycollate broth
- Size:125 ml
- Closure type:Flip cap
- Filling:100 ml



