Media in Bottles
Vegetable Peptone Broth, —, 10000 ml, Without cap, -, 1
Supplier: MERCK LIFE SCIENCE
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:1
- Standards:—
- Description:Vegetable Peptone Broth
- Size:-
- Closure type:Without cap
- Filling:10000 ml



