Digital block heater, compact
Product Details & Documents
These versatile digital block heaters provide good temperature uniformity and stability ensuring reproducible results. Units are compact, light weight with digital temperature controls and display, they maximises valuable bench space. The units are ideal for sample preparation, enzyme preservation, DNA amplification and enzyme–substrate reactions.
- Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
- Designed for precise temperature control
- Fast heating rate
- Multiple combination of different size of blocks allows for maximum tube size versatility during experimental procedure
- 12V low voltage power input
Delivery information: Supplied with block extraction tool and plastic lid, but without blocks, these must be ordered separately.
Caution: Temp. stability value shown above is at 37 °C.
* RT = Ambient
** S - 1 block, 2×½ blocks, 4×¼ blocks or 1×½ block plus 2×¼ blocks
*** D - 2 blocks, 4×½ blocks, 8×¼ blocks or combination
Specifications
- Heating capacity:58 W
- No. of blocks:1
- Temperature Range:RT* +2...100 °C
- Weight:1 kg
- Seq:01
- W×D×H:185×185×25 mm
- Temp. stability:≤±0,5 °C
- Type:Compact Dry Bath S**
- Cat. No.:460-0284
- Temperature stability:≤±0,5 °C
Accessories
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- ½ aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- ¼ aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- ¼ aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- ½ aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Plastic lid
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Plastic lid
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Heat insulator
Product Information
- Catalog #ForW×D×HAvailabilityPrice
- Full aluminium blockFull aluminium block460-031996-well microplate108×72×18 mmLog in for availability1 * 1 ST - 108,00 €
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Heat insulator
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- ¼ aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- ¼ aluminium block
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



