Digital block heaters
Product Details & Documents
Versatile digital block heaters offer a wide range of blocks to accommodate various tubes provide temperature uniformity and stability for reproducible results. The units have digital controls and display of time and temperature and are durable with a powder-coated steel body. Ideal for sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation. Choice of models to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
- Built-in temperature sensing probe for outstanding temperature accuracy and control
- Precise temperature control with PID circuit
- Temperature calibration enables user to offset the temperature to desired value
- Timer allows the user to accurately monitor the heating time
- Interchangeable aluminum alloy sample blocks provide versatility and allow for easy cleaning and disinfecting
Delivery information: Supplied with block extraction tool, but without blocks, these must be ordered separately.
Caution: * RT = Ambient
Note: Temp. stability value shown above is at 37 °C.
Specifications
- Heating capacity:600 W
- No. of blocks:4
- Temperature Range:RT* +5...130 °C
- Weight:4,7 kg
- W×D×H:450×200×100 mm
- Temp. stability:≤±1 °C
- Type:Block heater, digital
Accessories
Product Information
- Catalog #AvailabilityPrice
- PT1000 temperature probe
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium blockAluminium block460-029825 mm + 12-13 mm + 6 mm Ø tubes3+12+6Log in for availability1 * 1 ST - 172,00 €
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Mixed aluminium block
Product Information
- Catalog #ForW×D×HAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium blockAluminium block460-03051×96 well PCR plate, half or full skirted-Log in for availability1 * 1 ST - 266,00 €
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium blockAluminium block460-03041×96-well PCR plate, non skirted-Log in for availability1 * 1 ST - 232,00 €
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



