Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Round handled needle holders

Clamping length of 7 mm, Lockable, Stainless steel, 125 mm, Straight, 0,6 mm
  FSCI12075-12
Marketsource
Round handled needle holders
  FSCI12075-12
Marketsource
Round handled needle holders
Round handled needle holders

Clamping length of 7 mm, Lockable, Stainless steel, 125 mm, Straight, 0,6 mm

 
278,00 €
   
1 * 1 ST
 
 
  • Type:
    Lockable
  • Description:
    Clamping length of 7 mm
  • Length:
    125 mm
  • Material:
    Stainless steel
  • Seq:
    01
  • Tip width:
    0,6 mm
  • Tip shape:
    Straight