Feeder cabinets
Feeder cabinets, front load and front dispense with 4 compartments, 500×400×2000 mm
Catalog # PALBFFCS-0540-SST
Supplier: Palbam
Jump to:
Product Details & Documents
The feeder cabinets support the gowning operation. Made of stainless steel 304 with electropolished finish.
- Front access doors are used for reloading and for storage purposes
- The bottom cabinet can be used for storage or disposal
- Perspex dispenser slots ensure that garments/disposables can be extracted one at a time and do not spill out
Delivery information: Typically available from stock for direct shipment from the supplier. Delivered as shipped assembled or flat packed.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifications
- Description:Feeder cabinets, front load and front dispense with 4 compartments
- Seq:01
- W×D×H:500×400×2000 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.