Orders LinkContactUsLinkComponent
Accessories
Accessories
Cleaning tool (one tube holder, three tubes with caps) for use T 25 digital, T 25 easy clean digital/control homogenisers. Recommended cleaning process is: Pre-clean with suitable solvent, clean with detergent and final clean and rinse with demineralised water
Catalog # IKAA20003013
Supplier:   IKA
Accessories
Catalog # IKAA20003013
Supplier:  IKA
Accessories
Cleaning tool (one tube holder, three tubes with caps) for use T 25 digital, T 25 easy clean digital/control homogenisers. Recommended cleaning process is: Pre-clean with suitable solvent, clean with detergent and final clean and rinse with demineralised water
Price
126,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Cleaning tool (one tube holder, three tubes with caps) for use T 25 digital, T 25 easy clean digital/control homogenisers. Recommended cleaning process is: Pre-clean with suitable solvent, clean with detergent and final clean and rinse with demineralised water