Media in Bottles
TSA Tryptic soy agar, —, 100 ml, Without cap, 125 ml, 100
Catalog # HEIP1464290100
Supplier: MERCK LIFE SCIENCE
Jump to:
Product Details & Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:100
- Standards:—
- Description:TSA Tryptic soy agar
- Size:125 ml
- Closure type:Without cap
- Filling:100 ml



