Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, pH 4,00

Buffer solutions, pH 4,00

Buffer solution, 4,00, ±0,02, 20 °C, 10 L
Catalog # APPC272168.1315
Supplier:   PanReac AppliChem
Buffer solutions, pH 4,00
Catalog # APPC272168.1315
Supplier:  PanReac AppliChem
CAS Number:  
Buffer solutions, pH 4,00
Buffer solutions, pH 4,00

Buffer solution, 4,00, ±0,02, 20 °C, 10 L

CAS Number:  
Price
177,00 €
Log in for availability
1 * 10 L
Product Details & Documents

Buffer solutions contain citric acid 1 H₂O (11,768 g); sodium hydroxide 50% (6 ml); hydrochlorid acid 35% (3,96 ml); preservative 1% (5 ml) and water (1 l).

  • pH (25 to 40 °C): 4,00 +0,01
  • pH (20 °C): 4,00 ±0,02
  • pH (15 °C): 4,00 +0,01
  • pH (10 °C): 4,00 +0,02
  • pH (5 °C): 4,00 +0,04
  • pH (0 °C): 4,00 +0,05

Certifications: NIIST traceable.

Documents

Specifications

Specifications

  • Pk:
    10 L
  • Reference temperature:
    20 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,02
  • pH value:
    4,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.