Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions, pH 4,00

Buffer solution, 4,00, ±0,02, 20 °C, 250 mL
Catalog # APPC272168.1209
Supplier:   PanReac AppliChem
Buffer solutions, pH 4,00
Catalog # APPC272168.1209
Supplier:  PanReac AppliChem
CAS Number:  
Buffer solutions, pH 4,00
Buffer solutions, pH 4,00

Buffer solution, 4,00, ±0,02, 20 °C, 250 mL

CAS Number:  
Price
13,60 €
Log in for availability
1 * 250 mL
Product Details & Documents

Buffer solutions contain citric acid 1 H₂O (11,768 g); sodium hydroxide 50% (6 ml); hydrochlorid acid 35% (3,96 ml); preservative 1% (5 ml) and water (1 l).

  • pH (25 to 40 °C): 4,00 +0,01
  • pH (20 °C): 4,00 ±0,02
  • pH (15 °C): 4,00 +0,01
  • pH (10 °C): 4,00 +0,02
  • pH (5 °C): 4,00 +0,04
  • pH (0 °C): 4,00 +0,05

Certifications: NIIST traceable.

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Reference temperature:
    20 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,02
  • pH value:
    4,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.