Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
ME 10, 0,5 - 10 µl, 0,1 µl, ≤ ±1,5 - ±4,0 %, ≤ ±1,5 - ±4,0 %
Catalog # WITG5.403.201
Supplier: witeg Labortechnik
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
Catalog # WITG5.403.201
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
These multi-channel pipettes are designed for comfort over long term repetitive pipetting. 8 or 12 identical volumes for titration plates.
- Highest possible accuracy and precision
- Easy readable digital volume display
- Volume ranges colour coded on the push button
- Volumes steplessly adjustable
- Tip ejector can be blocked to avoid mis-application when using the instrument over large series
Delivery information: Each pipette is individually tested and delivered with a test certificate.
Specifications
- Volume:0,5 - 10 µl
- Description:ME 10
- Seq:04
- Increment:0,1 µl
- Accuracy:≤ ±1,5 - ±4,0 %
- Imprecision:≤ ±1,5 - ±4,0 %
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.