Order Entry
Belgium
Orders LinkContactUsLinkComponent
MPI-5a ≥95%

MPI-5a ≥95%

10 mg
Catalog # CAYM15200-10
Supplier:   Cayman Chemical
MPI-5a ≥95%
Catalog # CAYM15200-10
Supplier:  Cayman Chemical
CAS Number:  1259296-46-2
MPI-5a ≥95%

10 mg

CAS Number:  1259296-46-2
Price
1 290,00 €
Log in for availability
1 * 10 mg
Product Details & Documents

HDAC6 Inhibitor is a potent and selective inhibitor of HDAC6 (IC50= 6 nM) that poorly blocks other HDAC enzymes.

HDAC6 is a predominantly cytoplasmic enzyme that targets α-tubulin, cortactin, and heat shock protein 90, as well as other substrates. In this way, it impacts development, proliferation, invasion, and tumorigenesis. It is cell permeable, inhibiting the acetylation of tubulin in cells with an IC50 value of 210 nM.

Synonyms:  N-Hydroxy-2-(1-methyl-1H-pyrrole-2-carbonyl)-1,2,3,4-tetrahydroisoquinoline-6-carboxamide, MPI_5a
  • CAS Number:1259296-46-2
  • Formula:C₁₆H₁₇N₃O₃
  • MDL Number:MFCD29770783
  • Molecular Weight:299.32 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.