Isolator cleaning tool
Product Details & Documents
Isolator cleaning tool: Mop hardware, replacement head, replacement handle.
Specially engineered to work with isolators and other small areas, this easy to manoeuvre tool offers lightweight, one handed operation. With a low profile, swivel head, the isolator cleaning tool gets into 'hard to reach' places.
Engineered for quick cover changes during operation, the padded head allows for a consistent mop across different surfaces without losing cleaning power. The rounded edges clean the isolator corners and prevent scratches or damage to surfaces.
- Rapid change system allows user to replace covers/pads frequently, adhering to strict cleaning protocols
- Polyester pad conforms to surfaces
- Gets into 'hard to reach' areas with its low profile, swivel head
- 100% polyester covers provide the cleanest substrates available for cleaning isolators
- Offers one handed operation with lightweight handles
Isolator cleaning covers:
Made from 100% polyester or 100% polyester microdenier fabric. This fabric provides enhanced particle lifting, streak-free cleaning, plus enhanced sorption for working with solutions.
Enhanced absorbency and quick-wicking performance
Increased lifting and capturing of particles as small as microbes Streak-free cleaning.
Made from 100% polyester or 100% polyester microdenier fabric. This fabric provides enhanced particle lifting, streak-free cleaning, plus enhanced sorption for working with solutions.
Documents
Specifications
- Description:Isolator cleaning tool replacement head: with 1 thermoplastic mop head (100×180 mm), 1 pad
- Length:-
- L×W:100×180 mm
- Seq:2
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



