WASH BUFFER (20X
Catalog # MESOR61AA-1
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
HUMAN MCP-1/CCL2 QUANTIKINE ELISA KIT 1 * 96 Tests
Supplier: RD SYSTEMS EUROPE
Neoprene gloves, Microflex® NeoTouch® 25-101 / 25-201
Supplier: Ansell
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
DILUENT 11, 50ML 1 * 50 mL
Supplier: MESO SCALE DISCOVERY
MSD GOLD Read Buffer B 1 * 90 mL
Supplier: MESO SCALE DISCOVERY
APPLIED BIOLOGICAL MATERIALS|PRIGROW I M 1 * 500 mL
Supplier: MARKET SOURCE PART PROCESS
CONTROL W. COVER LCTR 6+ CY509109 NR 1 * 1 items
Supplier: Thermo Fisher Scientific
R-PLEX Human Osteoactivin Assay 5 plate 1 * 1 KIT
Supplier: MESO SCALE DISCOVERY
MAGPIX CALIBRATION KIT 1 * 1 KIT
Supplier: MARKET SOURCE PART PROCESS
VWR®, Replacement LED for Magnifiers
Supplier: VWR Collection