FUSIDIC ACID IMPURITY MIXTURE 1 * 0,47 mg
Catalog # EDQMY0001384
Supplier: EDQM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Syringe filters, Minisart® RC
Supplier: Sartorius
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
FUSIDIC ACID FOR PEAK IDENTIFICATION 1 * 5 mg
Supplier: EDQM