NORADRENALINE IMPURITY F 1 * 10 mg
Catalog # EDQMY0000686
Supplier: EDQM
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Supplier: HAMILTON BONADUZ
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Sulphuric acid 1 mol/l (2 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals